2-(4-Methanesulfonylpiperazin-1-yl)ethan-1-ol ≥95%
Supplier: Matrix Scientific
056416-1G
056416-500MG
056416-5G
101910-428EA
710.77
USD
101910-428
101910-426
101910-430
2-(4-Methanesulfonylpiperazin-1-yl)ethan-1-ol ≥95%
2-(4-Methanesulfonylpiperazin-1-yl)ethan-1-ol
Hazard: Irritant
Formula:
C₇H₁₆N₂O₃S |
MDL Number:
MFCD01212002 CAS Number: 72388-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...