Ethyl-N-[2-(4-hydroxyphenyl)ethyl]carbamate ≥95%
Supplier: Matrix Scientific
056402-500MG
056402-1G
101910-364EA
698.46
USD
101910-364
101910-362
Ethyl-N-[2-(4-hydroxyphenyl)ethyl]carbamate ≥95%
Ethyl-N-[2-(4-hydroxyphenyl)ethyl]carbamate
Hazard: Irritant
Formula:
C₁₁H₁₅NO₃ |
MDL Number:
MFCD02682912 CAS Number: 70275-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...