1,5-Dimethyl-1H-pyrrole-2-carboximidamide hydrochloride ≥95%

Supplier: Matrix Scientific

056398-1G 056398-500MG
101910-348EA 698.46 USD
101910-348 101910-346
1,5-Dimethyl-1H-pyrrole-2-carboximidamide hydrochloride ≥95%
1,5-Dimethyl-1H-pyrrole-2-carboximidamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₂ClN₃
MDL Number: MFCD11052487
CAS Number: 744193-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR