6-(tert-Butyl)-1-ethyl 6-azaspiro[2.5]octane-1,6-dicarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
6-tert-Butyl 1-ethyl 6-azaspiro[2.5]octane-1,6-dicarboxylate
056365-1G
056365-500MG
101910-214EA
467.69
USD
101910-214
101910-212
6-(tert-Butyl)-1-ethyl 6-azaspiro[2.5]octane-1,6-dicarboxylate ≥95%
6-(tert-Butyl)-1-ethyl 6-azaspiro[2.5]octane-1,6-dicarboxylate
Hazard: Irritant
Formula:
C₁₅H₂₅NO₄ MW: 283.37 g/mol |
MDL Number:
MFCD16622771 CAS Number: 1242268-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...