tert-Butyl-6-aminopyridin-2-ylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6-aminopyridin-2-ylcarbamate2-Amino-6-(tert-butoxycarbonylamino)pyridine2-Amino-6-(Boc-amino)pyridine

056362-1G 056362-500MG
101910-202EA 93.85 USD
101910-202 101910-200
tert-Butyl-6-aminopyridin-2-ylcarbamate ≥95%
tert-Butyl-6-aminopyridin-2-ylcarbamate

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂
MW: 209.2500 g/mol
MDL Number: MFCD09037865
CAS Number: 322690-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR