6-(1H-Pyrrol-1-yl)-1,3-benzodioxol-5-amine ≥95%

Supplier: Matrix Scientific

056360-500MG 056360-1G
101910-194EA 1163.08 USD
101910-194 101910-192
6-(1H-Pyrrol-1-yl)-1,3-benzodioxol-5-amine ≥95%
6-(1H-Pyrrol-1-yl)-1,3-benzodioxol-5-amine

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD16622819
CAS Number: 1242268-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR