Methyl-2-cyano-2-(5-cyano-2-pyridinyl)acetate ≥95%
Supplier: Matrix Scientific
056355-1G
056355-500MG
101910-174EA
278.46
USD
101910-174
101910-176
Methyl-2-cyano-2-(5-cyano-2-pyridinyl)acetate ≥95%
Methyl-2-cyano-2-(5-cyano-2-pyridinyl)acetate
Hazard: Irritant
Formula:
C₁₀H₇N₃O₂ MW: 201.19 g/mol |
MDL Number:
MFCD16622815 CAS Number: 1242268-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...