2-Methoxynicotinohydrazide ≥95%

Supplier: Matrix Scientific

056352-1G 056352-500MG
101910-162EA 350.77 USD
101910-162 101910-160
2-Methoxynicotinohydrazide ≥95%
2-Methoxynicotinohydrazide

Hazard: Irritant

Formula: C₇H₉N₃O₂
MDL Number: MFCD11644032
CAS Number: 89853-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR