2-Isopropylpiperazine ≥95%

Supplier: Matrix Scientific

056351-1G 056351-500MG
101910-158EA 483.08 USD
101910-158 101910-156
2-Isopropylpiperazine ≥95%
2-Isopropylpiperazine

Hazard: Irritant

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
MDL Number: MFCD07373042
CAS Number: 84468-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR