5-[(E)-(4-Hydroxyphenyl)methylidene]-1,3-thiazolane-2,4-dione ≥95%
Supplier: Matrix Scientific
056350-500MG
056350-1G
056350-5G
101910-152EA
232.31
USD
101910-152
101910-150
101910-154
5-[(E)-(4-Hydroxyphenyl)methylidene]-1,3-thiazolane-2,4-dione ≥95%
5-[(E)-(4-Hydroxyphenyl)methylidene]-1,3-thiazolane-2,4-dione
Hazard: Irritant
Formula:
C₁₀H₇NO₃S |
MDL Number:
MFCD00640639 CAS Number: 103788-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...