3-(Hexyloxy)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

056348-500MG 056348-1G 056348-5G
101910-140EA 181.54 USD
101910-140 101910-144 101910-142
3-(Hexyloxy)benzenecarbonitrile ≥95%
3-(Hexyloxy)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₁₇NO
MDL Number: MFCD09710699
CAS Number: 910452-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR