4-Fluoro-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

056346-1G
101910-134EA 135.38 USD
101910-134
4-Fluoro-2-methoxypyridine ≥95%
4-Fluoro-2-methoxypyridine

Hazard: Irritant

Formula: C₆H₆FNO
MDL Number: MFCD09835267
CAS Number: 96530-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR