Ethyl-2-cyclohexylidenacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-cyclohexylideneacetate

056341-500MG 056341-1G
101910-112EA 350.77 USD
101910-112 101910-110
Ethyl-2-cyclohexylidenacetate ≥95%
Ethyl-2-cyclohexylidenacetate

Hazard: Irritant

Formula: C₁₀H₁₆O₂
MW: 168.24 g/mol
MDL Number: MFCD00963857
CAS Number: 1552-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR