5H-Spiro[[1,3]dioxolo[4,5-g]pyrrolo[1,2-a]quinoxaline-4,4'-piperidine] hydrochloride ≥95%
Supplier: Matrix Scientific
056338-1G
056338-500MG
101910-100EA
1400
USD
101910-100
101910-098
5H-Spiro[[1,3]dioxolo[4,5-g]pyrrolo[1,2-a]quinoxaline-4,4'-piperidine] hydrochloride ≥95%
5H-Spiro[[1,3]dioxolo[4,5-g]pyrrolo[1,2-a]quinoxaline-4,4'-piperidine] hydrochloride
Hazard: Irritant
Formula:
C₁₆H₁₈ClN₃O₂ |
MDL Number:
MFCD16622820 CAS Number: 1242267-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...