2,4-Dichloroquinazoline ≥95%

Supplier: Matrix Scientific

056334-5G
101910-082EA 24.15 USD
101910-082
2,4-Dichloroquinazoline ≥95%
2,4-Dichloroquinazoline

Hazard: Irritant

Formula: C₈H₄Cl₂N₂
Melting Pt: 117…119 °C
MDL Number: MFCD00091950
CAS Number: 607-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR