1-Cyanocyclopropanecarbohydrazide ≥95%

Supplier: Matrix Scientific

056332-500MG 056332-1G
101910-074EA 698.46 USD
101910-074 101910-072
1-Cyanocyclopropanecarbohydrazide ≥95%
1-Cyanocyclopropanecarbohydrazide

Hazard: Irritant

Formula: C₅H₇N₃O
MDL Number: MFCD03549791
CAS Number: 401825-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR