3-Chloroimidazo[1,2-a]pyrimidine ≥95%

Supplier: Matrix Scientific

056324-500MG 056324-1G
101910-038EA 467.69 USD
101910-038 101910-036
3-Chloroimidazo[1,2-a]pyrimidine ≥95%
3-Chloroimidazo[1,2-a]pyrimidine

Hazard: Irritant

Formula: C₆H₄ClN₃
Melting Pt: 158…160 °C
MDL Number: MFCD09994773
CAS Number: 143696-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR