3-Chloro-5,7-dihydroxy-4-methyl-2H-chromen-2-one ≥95%

Supplier: Matrix Scientific

056321-500MG 056321-1G
101910-026EA 256.92 USD
101910-026 101910-024
3-Chloro-5,7-dihydroxy-4-methyl-2H-chromen-2-one ≥95%
3-Chloro-5,7-dihydroxy-4-methyl-2H-chromen-2-one

Hazard: Irritant

Formula: C₁₀H₇ClO₄
MDL Number: MFCD00488345
CAS Number: 22649-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR