3-Bromo-2-pyridinecarbohydrazide ≥95%

Supplier: Matrix Scientific

056316-500MG
101910-006EA 556.92 USD
101910-006
3-Bromo-2-pyridinecarbohydrazide ≥95%
3-Bromo-2-pyridinecarbohydrazide

Hazard: Irritant

Formula: C₆H₆BrN₃O
MDL Number: MFCD16622811
CAS Number: 1242267-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR