5-Bromo-2-chloroisonicotinamide ≥95%

Supplier: Matrix Scientific

056312-500MG 056312-1G
101909-992EA 416.3 USD
101909-992 101909-994
5-Bromo-2-chloroisonicotinamide ≥95%
5-Bromo-2-chloroisonicotinamide

Hazard: Irritant

Formula: C₆H₄BrClN₂O
MDL Number: MFCD16622808
CAS Number: 1242268-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR