N-(5-Amino-3-chloro-2-pyridinyl)-N-methylamine ≥95%

Supplier: Matrix Scientific

056308-500MG
101909-976EA 921.54 USD
101909-976
N-(5-Amino-3-chloro-2-pyridinyl)-N-methylamine ≥95%
N-(5-Amino-3-chloro-2-pyridinyl)-N-methylamine

Hazard: Irritant

Formula: C₆H₈ClN₃
MDL Number: MFCD16622774
CAS Number: 813424-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR