Methyl-2-amino-3,4-difluorobenzoate ≥97%
Supplier: Matrix Scientific
056304-1G
056304-5G
101909-970EA
320
USD
101909-970
101909-968
Methyl-2-amino-3,4-difluorobenzoate ≥97%
Methyl 2-amino-3,4-difluorobenzoate
Hazard: Irritant
Formula:
C₈H₇F₂NO₂ |
MDL Number:
MFCD09996886 CAS Number: 170108-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...