4-Chloro-2,3,5,6-tetrafluoropyridine ≥98%

Supplier: Matrix Scientific

056291-1G 056291-5G
101909-950EA 80 USD
101909-950 101909-952
4-Chloro-2,3,5,6-tetrafluoropyridine ≥98%
4-Chloro-2,3,5,6-tetrafluoropyridine

Hazard: Irritant

Formula: C₅ClF₄N
MDL Number: MFCD00971927
CAS Number: 52026-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR