1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carboxylic acid

Supplier: Matrix Scientific

055674-100MG
101909-904EA 244.62 USD
101909-904
1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carboxylic acid
1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₄
MDL Number: MFCD16628275
CAS Number: 1228665-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR