1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carbonitrile

Supplier: Matrix Scientific

055673-100MG
101909-902EA 364.62 USD
101909-902
1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carbonitrile
1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carbonitrile

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₂
MDL Number: MFCD16628274
CAS Number: 1228666-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR