(E)-1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carbaldehyde oxime

Supplier: Matrix Scientific

055672-100MG
101909-900EA 364.62 USD
101909-900
(E)-1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carbaldehyde oxime
(E)-1-Pivaloyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine-6-carbaldehyde oxime

Hazard: Irritant

Formula: C₁₃H₁₇N₃O₃
MDL Number: MFCD16628273
CAS Number: 1228670-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR