2,2-Dimethyl-1-(6-((trimethylsilyl)ethynyl)-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazin-1-yl)propan-1-on

Supplier: Matrix Scientific

055668-100MG
101909-892EA 364.62 USD
101909-892
2,2-Dimethyl-1-(6-((trimethylsilyl)ethynyl)-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazin-1-yl)propan-1-on
2,2-Dimethyl-1-(6-((trimethylsilyl)ethynyl)-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazin-1-yl)propan-1-on

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₂Si
MDL Number: MFCD16628269
CAS Number: 1228666-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR