2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine-8-carbonitrile

Supplier: Matrix Scientific

055663-100MG
101909-882EA 246.15 USD
101909-882
2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine-8-carbonitrile
2,3-Dihydro-1H-pyrido[3,4-b][1,4]oxazine-8-carbonitrile

Hazard: Irritant

Formula: C₈H₇N₃O
MDL Number: MFCD15530319
CAS Number: 1228666-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR