1-(5-Fluoro-1H-pyrrolo[2,3-b]pyridin-4-yl)ethanone

Supplier: Matrix Scientific

055648-100MG
101909-852EA 364.62 USD
101909-852
1-(5-Fluoro-1H-pyrrolo[2,3-b]pyridin-4-yl)ethanone
1-(5-Fluoro-1H-pyrrolo[2,3-b]pyridin-4-yl)ethanone

Hazard: Irritant

Formula: C₉H₇FN₂O
MDL Number: MFCD16628259
CAS Number: 1228666-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR