5-Bromo-2-ethyl-3-iodo-1H-pyrrolo[2,3-b]pyridine
Supplier: Matrix Scientific
055644-100MG
101909-844EA
364.62
USD
101909-844
5-Bromo-2-ethyl-3-iodo-1H-pyrrolo[2,3-b]pyridine
5-Bromo-2-ethyl-3-iodo-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₉H₈BrIN₂ |
MDL Number:
MFCD16628255 CAS Number: 1228666-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...