5-Fluoro-4-methyl-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

055643-100MG
101909-842EA 244.62 USD
101909-842
5-Fluoro-4-methyl-1H-pyrrolo[2,3-b]pyridine
5-Fluoro-4-methyl-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₈H₇FN₂
MDL Number: MFCD16628254
CAS Number: 1228666-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR