5-Fluoro-1H-pyrrolo[2,3-b]pyridine-4-carbaldehyde

055642-100MG
101909-840EA 1997.34 USD
101909-840
5-Fluoro-1H-pyrrolo[2,3-b]pyridine-4-carbaldehyde
5-Fluoro-1H-pyrrolo[2,3-b]pyridine-4-carbaldehyde

Hazard: Irritant

Formula: C₈H₅FN₂O
MDL Number: MFCD12962822
CAS Number: 1190310-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR