4-Allyl-5-fluoro-1H-pyrrolo[2,3-b]pyridine
Supplier: Matrix Scientific
055636-100MG
101909-828EA
364.62
USD
101909-828
4-Allyl-5-fluoro-1H-pyrrolo[2,3-b]pyridine
4-Allyl-5-fluoro-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₁₀H₉FN₂ |
MDL Number:
MFCD16628248 CAS Number: 1228665-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...