5-Bromo-2-ethyl-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

055633-100MG
101909-822EA 246.15 USD
101909-822
5-Bromo-2-ethyl-1H-pyrrolo[2,3-b]pyridine
5-Bromo-2-ethyl-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₉H₉BrN₂
MDL Number: MFCD16628246
CAS Number: 1228666-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR