5-(Trifluoromethyl)-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid

Supplier: Matrix Scientific

055632-100MG
101909-820EA 364.62 USD
101909-820
5-(Trifluoromethyl)-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid
5-(Trifluoromethyl)-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₅F₃N₂O₂Si
MDL Number: MFCD15530313
CAS Number: 1222533-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR