1-(5-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)ethanone
Supplier: Matrix Scientific
055629-100MG
101909-814EA
364.62
USD
101909-814
1-(5-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)ethanone
1-(5-Methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)ethanone
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O |
MDL Number:
MFCD16628244 CAS Number: 1222533-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...