2,2-Dimethyl-1-(6-((trimethylsilyl)ethynyl)-3,4-dihydro-1,8-naphthyridin-1(2H)-yl)propan-1-one

Supplier: Matrix Scientific

055622-100MG
101909-800EA 364.62 USD
101909-800
2,2-Dimethyl-1-(6-((trimethylsilyl)ethynyl)-3,4-dihydro-1,8-naphthyridin-1(2H)-yl)propan-1-one
2,2-Dimethyl-1-(6-((trimethylsilyl)ethynyl)-3,4-dihydro-1,8-naphthyridin-1(2H)-yl)propan-1-one

Hazard: Irritant

Formula: C₁₈H₂₆N₂OSi
MDL Number: MFCD16628238
CAS Number: 1222533-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR