6-Fluoro-3,4-dihydro-1,8-naphthyridin-2(1H)-one

055620-100MG
101909-796EA 782.12 USD
101909-796
6-Fluoro-3,4-dihydro-1,8-naphthyridin-2(1H)-one
6-Fluoro-3,4-dihydro-1,8-naphthyridin-2(1H)-one

Hazard: Irritant

Formula: C₈H₇FN₂O
Melting Pt: 226 °C
MDL Number: MFCD16628236
CAS Number: 1222533-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR