6-Fluoro-2-methyl-1,8-naphthyridine

Supplier: Matrix Scientific

055616-100MG
101909-788EA 364.62 USD
101909-788
6-Fluoro-2-methyl-1,8-naphthyridine
6-Fluoro-2-methyl-1,8-naphthyridine

Hazard: Irritant

Formula: C₉H₇FN₂
MDL Number: MFCD16628232
CAS Number: 1222533-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR