2-Methoxy-6-(pyrrolidin-1-yl)nicotinic acid

Supplier: Matrix Scientific

055612-100MG
101909-780EA 244.62 USD
101909-780
2-Methoxy-6-(pyrrolidin-1-yl)nicotinic acid
2-Methoxy-6-(pyrrolidin-1-yl)nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃
MDL Number: MFCD15530310
CAS Number: 1228665-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR