2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-6-fluoro-4-iodopyridine
Supplier: Matrix Scientific
055610-100MG
101909-776EA
364.62
USD
101909-776
2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-6-fluoro-4-iodopyridine
2-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-6-fluoro-4-iodopyridine
Hazard: Irritant
Formula:
C₁₆H₂₆FIN₂OSi |
MDL Number:
MFCD15530308 CAS Number: 1228665-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...