3-Allyl-2-methoxy-6-(pyrrolidin-1-yl)pyridine

Supplier: Matrix Scientific

055609-100MG
101909-774EA 364.62 USD
101909-774
3-Allyl-2-methoxy-6-(pyrrolidin-1-yl)pyridine
3-Allyl-2-methoxy-6-(pyrrolidin-1-yl)pyridine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MDL Number: MFCD15530307
CAS Number: 1228666-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR