2-Fluoro-3,4-diiodo-6-(pyrrolidin-1-yl)pyridine

Supplier: Matrix Scientific

055608-100MG
101909-772EA 364.62 USD
101909-772
2-Fluoro-3,4-diiodo-6-(pyrrolidin-1-yl)pyridine
2-Fluoro-3,4-diiodo-6-(pyrrolidin-1-yl)pyridine

Hazard: Irritant

Formula: C₉H₉FI₂N₂
MDL Number: MFCD15530306
CAS Number: 1228665-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR