2-Methoxy-6-(pyrrolidin-1-yl)nicotinaldehyde

Supplier: Matrix Scientific

055604-100MG
101909-764EA 364.62 USD
101909-764
2-Methoxy-6-(pyrrolidin-1-yl)nicotinaldehyde
2-Methoxy-6-(pyrrolidin-1-yl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD15530302
CAS Number: 1228666-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR