(E)-Methyl-3-(4-chloropyridin-3-yl)acrylate

Supplier: Matrix Scientific

055599-100MG
101909-754EA 364.62 USD
101909-754
(E)-Methyl-3-(4-chloropyridin-3-yl)acrylate
(E)-Methyl-3-(4-chloropyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₉H₈ClNO₂
MDL Number: MFCD15530297
CAS Number: 1228670-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR