tert-Butyl-4-chloro-3-formylpyridin-2-ylcarbamate

Supplier: Matrix Scientific

Synonyms: tert-Butyl (4-chloro-3-formylpyridin-2-yl)carbamate

055596-100MG
101909-748EA 364.62 USD
101909-748
tert-Butyl-4-chloro-3-formylpyridin-2-ylcarbamate
tert-Butyl-4-chloro-3-formylpyridin-2-ylcarbamate

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃
MW: 256.69 g/mol
MDL Number: MFCD15530295
CAS Number: 868736-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR