3-(1-Benzylpyrrolidin-3-yl)-2-methoxy-5-methylpyridine

Supplier: Matrix Scientific

055593-100MG
101909-742EA 364.62 USD
101909-742
3-(1-Benzylpyrrolidin-3-yl)-2-methoxy-5-methylpyridine
3-(1-Benzylpyrrolidin-3-yl)-2-methoxy-5-methylpyridine

Hazard: Irritant

Formula: C₁₈H₂₂N₂O
MDL Number: MFCD15530292
CAS Number: 1228666-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR