(Z)-5-(Benzyloxy)-6-bromopicolinaldehyde oxime

Supplier: Matrix Scientific

055591-100MG
101909-738EA 364.62 USD
101909-738
(Z)-5-(Benzyloxy)-6-bromopicolinaldehyde oxime
(Z)-5-(Benzyloxy)-6-bromopicolinaldehyde oxime

Hazard: Irritant

Formula: C₁₃H₁₁BrN₂O₂
MDL Number: MFCD15530290
CAS Number: 1228670-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR