N-(3-(3-(tert-Butyldimethylsilyloxy)propyl)pyridin-2-yl)pivalamide

Supplier: Matrix Scientific

055590-100MG
101909-736EA 364.62 USD
101909-736
N-(3-(3-(tert-Butyldimethylsilyloxy)propyl)pyridin-2-yl)pivalamide
N-(3-(3-(tert-Butyldimethylsilyloxy)propyl)pyridin-2-yl)pivalamide

Hazard: Irritant

Formula: C₁₉H₃₄N₂O₂Si
MW: 350.58 g/mol
MDL Number: MFCD15530289
CAS Number: 1228665-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR