6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoro-3-iodopyridine

Supplier: Matrix Scientific

055584-100MG
101909-724EA 364.62 USD
101909-724
6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoro-3-iodopyridine
6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoro-3-iodopyridine

Hazard: Irritant

Formula: C₁₆H₂₆FIN₂OSi
MW: 436.39 g/mol
MDL Number: MFCD15530283
CAS Number: 1228666-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR